Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EIF2B5.
Supplier: Abnova Corporation H00008893A01
Description
Sequence: LPLLKAWSPVFRNYIKRAADHLEALAAIEDFFLEHEALGISMAKVLMAFYQLEILAEETILSWFSQRDTTDKGQQLRKNQQLQRFIQWLKEAEEESSEDSpecifications
eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant EIF2B5. | |
EIF2B5 | |
CACH/CLE/EIF-2B/EIF2Bepsilon/LVWM | |
Mouse | |
50 μL | |
Yes | |
8893 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_003907 | |
EIF2B5 | |
EIF2B5 (NP_003898, 622 a.a. ∼ 720 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction