Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF2D, Mouse, Clone: 2D10, Abnova™

Catalog No. 89021397 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-397 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-397 Supplier Abnova Corporation Supplier No. H00001939M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant EIF2D.

This gene encodes a protein receptor that localizes phosphoglycoproteins within endosomes and at the cell periphery. This trafficking receptor for phosphoglycoproteins may play a role in neuroplasticity by modulating cell-cell interactions, intracellular adhesion, and protein binding at membrane surfaces. In hippocampal neurons, long-lasting down-regulation of ligatin mRNA levels occurs via post-transcriptional RNA processing following glutamate receptor activation. This protein contains single PUA and SUI1 domains and these domains may function in RNA binding and translation initiation, respectively. [provided by RefSeq

Sequence: KGRICPIDITLAQRASNKKVTVVRNLEAYGLDPYSVAAILQQRCQASTTVNPAPGAKDSLQVQIQGNQVHHLGWLLLEEYQLPRKHIQGLEKALKPGKKK

Specifications

Antigen EIF2D
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant EIF2D.
Formulation PBS with no preservative; pH 7.4
Gene EIF2D
Gene Accession No. NM_006893
Gene Alias LGTN; HCA56
Gene Symbols EIF2D
Host Species Mouse
Immunogen EIF2D (NP_008824.2, 485 a.a. ∼ 584 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1939
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.