Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eukaryotic translation initiation factor 4A, isoform 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EIF4A2.
Supplier: Abnova Corporation H00001974A01
Description
Sequence: MSGGSADYNREHGGPEGMDPDGVIESNWNEIVDNFDDMNLKESLLRGIYAYGFEKPSAIQQRAIIPCIKGYDVIAQAQSGTGKTATFAISILQQLEIEFKSpecifications
eukaryotic translation initiation factor 4A, isoform 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant EIF4A2. | |
EIF4A2 | |
BM-010/DDX2B/EIF4A/EIF4F | |
Mouse | |
50 μL | |
Yes | |
1974 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_001967 | |
EIF4A2 | |
EIF4A2 (NP_001958, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction