Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-eIF4A2, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$182.00

Specifications

Antigen eIF4A2
Concentration 0.5 mg/mL
Dilution Western Blot 0.5-1.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224612SS
SDP
View Documents
Novus Biologicals
NBP224612SS
0.025 mg
Each for $182.00
Only null left
Add to Cart
 
NBP224612
SDP
View Documents
Novus Biologicals
NBP224612
0.1 mg N/A N/A N/A
Description

Description

eIF4A2 Polyclonal specifically detects eIF4A2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

eIF4A2
Western Blot 0.5-1.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Polyclonal
Purified
RUO
Human, Mouse
Q14240
1974
A portion of amino acids 1-50 of human eIF4A2 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, mTOR Pathway
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
ATP-dependent RNA helicase eIF4A-2, BM-010, DDX2Beukaryotic translation initiation factor 4A, isoform 2, EC 3.6.1, EC 3.6.4.13, EIF4A, eIF-4A-II, eIF4A-II, EIF4F, eukaryotic initiation factor 4A-II, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A2
EIF4A2
IgG
Protein A purified
Human and mouse. 100% homologous in rat, bovine, and porcine.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.