Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF4A3 Mouse anti-Human, Mouse, Clone: 3 F 1, MilliporeSigma™

Catalog No. 051527MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-152-7MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-152-7MI Supplier MilliporeSigma Supplier No. 051527
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-eIF4A3 Antibody, clone 3F1 (mouse monoclonal antibody) validated in western blot, immunoprecipitation, immunocytochemistry to detect eIF4A3 also known as ATP-dependent RNA helicase DDX48, Eukaryotic translation initiation factor 4A isoform 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4A3
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3 F 1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_055555.1
Gene Symbols DDX48, EIF4AIII, NMP265, NUK-34, eIF4AIII, hNMP265
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human eIF4A3.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.