Learn More
EIF4A3, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human EIF4A3 peptide using ARM Technology.
Supplier: Abnova Corporation H00009775K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene is a nuclear matrix protein. Its amino acid sequence is highly similar to the amino acid sequences of the translation initiation factors eIF4AI and eIF4AII, two other members of the DEAD box protein family. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
EIF4A3 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
EIF4A3 | |
EIF4A3 | |
A synthetic peptide of human EIF4A3 | |
RUO | |
9775 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
eukaryotic translation initiation factor 4A, isoform 3 | |
EIF4A3 | |
DDX48, KIAA0111, MGC10862, NMP265, NUK-34, eIF4AIII, hNMP265 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.