Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF4G Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 071800MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-180-0MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-180-0MI Supplier MilliporeSigma Supplier No. 071800
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-eIF4G Antibody is an antibody against eIF4G for use in western blotting and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4G
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum with 0.05% NaN3.
Gene Symbols EIF4-gamma ; EIF4F; EIF4G ; EIF4GI; eIF-4G1; p220
Host Species Rabbit
Immunogen Synthetic peptide corresponding to N-terminal region of human eIF4G.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_937884
Target Species Human, Mouse
Content And Storage −20°C for up to 12 months
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.