Learn More
eukaryotic translation initiation factor 4 gamma, 3, Mouse, Clone: 1H5, Abnova™
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Antigen | eukaryotic translation initiation factor 4 gamma, 3 |
| Applications | ELISA, Immunohistochemistry (PFA fixed), KnockDown, Western Blot |
| Classification | Monoclonal |
| Clone | 1H5 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant EIF4G3. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | EIF4G3 |
| Gene Accession No. | BC030578 |
| Gene Alias | eIF4GII |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.