Learn More
elastase 3A, pancreatic, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human ELA3A protein.
Supplier: Abnova Corporation H00010136D01P
Description
Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Unlike other elastases, elastase 3A has little elastolytic activity. Like most of the human elastases, elastase 3A is secreted from the pancreas as a zymogen and, like other serine proteases such as trypsin, chymotrypsin and kallikrein, it has a digestive function in the intestine. Elastase 3A preferentially cleaves proteins after alanine residues. Elastase 3A may also function in the intestinal transport and metabolism of cholesterol. Both elastase 3A and elastase 3B have been referred to as protease E and as elastase 1. [provided by RefSeq
Sequence: MMLRLLSSLLLVAVASGYGPPSSHSSSRVVHGEDAVPYSWPWQVSLQYEKSGSFYHTCGGSLIAPDWVVTAGHCISRDLTYQVVLGEYNLAVKEGPEQVIPINSEELFVHPLWNRSCVACGNDIALIKLSRSAQLGDAVQLASLPPAGDILPNKTPCYITGWGRLYTNGPLPDKLQQARLPVVDYKHCSRWNWWGSTVKKTMVCAGGYIRSGCNGDSGGPLNCPTEDGGWQVHGVTSFVSGFGCNFIWKPTVFTRVSAFIDWIEETIASHSpecifications
elastase 3A, pancreatic | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human ELA3A protein. | |
ELA3A | |
ELA3 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005747.3 | |
ELA3A | |
ELA3A (NP_005738.3, 1 a.a. ∼ 270 a.a) full-length human protein. | |
100 μg | |
Endocrine/Metabolism | |
10136 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.