Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastin Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB2043MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2043MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2043MI Supplier MilliporeSigma Supplier No. AB2043
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Elastin using this Anti-Elastin Antibody validated for use in ELISA, RIA, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Elastin
Applications ELISA, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Formulation IgG fraction. Liquid in 0.01M Phosphate, 0.09M Sodium Chloride, pH 7.2, containing no preservatives.
Gene Symbols ELN; tropoelastin; FLJ38671; SVAS; WS; Tropoelastin; elastin; FLJ43523; WBS
Host Species Rabbit
Immunogen Elastin extracted and purified from human skin.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000501.2; NM_001081752.1; NM_001081753.1; NM_001081754.1; NM_001081755.1
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.