Learn More
E74-like factor 4 (ets domain transcription factor), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human ELF4 protein.
Supplier: Abnova Corporation H00002000D01P
Description
ELF4 is a member of the ETS (see MIM 164720) gene family of transcription factors (Aryee et al., 1998 [PubMed 9524226]). ELF4 is involved in inducing cell cycle arrest in naive CD8+ T cells (Yamada et al., 2009 [PubMed 19412182]).[supplied by OMIM
Sequence: MAITLQPSDLIFEFASNGMDDDIHQLEDPSVFPAVIVEQVPYPDLLHLYSGLELDDVHNGIITDGTLCMTQDQILEGSFLLTDDNEATSHTMSTAEVLLNMESPSDILDEKQIFSTSEMLPDSDPAPAVTLPNYLFPASEPDALNRAGDTSDQEGHSLEEKASREESAKKTGKSKKRIRKTKGNRSTSPVTDPSIPIRKKSKDGKGSTIYLWEFLLALLQDRNTCPKYIKWTQREKGIFKLVDSKAVSKLWGKQKNKPDMNYETMGRALRYYYQRGILAKVEGQRLVYQFKEMPKDLVVIEDEDESSEATAAPPQASTASVASASTTRRTSSRVSSRSAPQGKGSSSWEKPKIQHVGLQPSASLELGPSLDEEIPTTSTMLVSPAEGQVKLTKAVSASSVPSNIHLGVAPVGSGSALTLQTIPLTTVLTNGPPASTTAPTQLVLQSVPAASTFKDTFTLQASFPLNASFQDSQVAAPGAPLILSGLPQLLAGANRPTNPAPPTVTGAGPAGPSSQPPGTVIAAFIRTSGTTAAPRVKEGPLRSSSYVQGMVTGAPMEGLLVPEETLRELLRDQAHLQPLPTQVVSRGSHNPSLLGNQTLSPPSRPTVGLTPVAELELSSGSGSLLMAEPSVTTSGSLLTRSPTPAPFSPFNPTSLIKMEPHDISpecifications
E74-like factor 4 (ets domain transcription factor) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human ELF4 protein. | |
ELF4 | |
ELFR/MEF | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001421.1 | |
ELF4 | |
ELF4 (NP_001412.1, 1 a.a. ∼ 663 a.a) full-length human protein. | |
100 μg | |
Transcription Regulation | |
2000 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.