Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

elF4E (pS209) Mouse, PE, Clone: J77-925, BD

Catalog No. BDB560229 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560229 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560229 Supplier BD Biosciences Supplier No. 560229

Mouse Monoclonal Antibody

The eukaryotic translation Initiation Factor 4E (eIF4E) is a 25kDa phosphoprotein that specifically binds to the 7-methylguanosine-containing cap of mRNA. eIF4E is the rate-limiting component for the initiation of cap-dependent translation by the eIF4F translation initiation complex, which is composed of eIF4E, eIF4G, and eIF4A. This complex promotes the unwinding of secondary structure at the 5′ untranslated region of mRNA that is necessary to expose and locate the AUG initiation codon. Other functions of eIF4E have been identified, such as promoting the export of mRNAs that are involved in cell cycle progression from the nucleus and differentially regulating the translation of certain mRNAs in the cytoplasm. Three mechanisms for eIF4E regulation have been identified: Mnk1-mediated phosphorylation on serine 209 (S209) is required for eIF4E binding to the cap structure; over-expression of phosphorylated eIF4E can lead to increased cell proliferation, suppression of apoptosis, and a transformed phenotype; and interactions with nonphosphorylated eIF4E-binding proteins inhibit the formation of the eIF4F complex. The J77-925 monoclonal antibody recognizes the phosphorylated S209 (pS209) of eIF4E.

Host Species: Mouse
Clone: J77-925
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Phosphorylated Human eIF4E Peptide

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen elF4E (pS209)
Applications Flow Cytometry
Classification Monoclonal
Clone J77-925
Conjugate PE
Description eIF-4E, CBP, EIF4E1, EIF4EL1, EIF4F, mRNA cap-binding protein
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human eIF4E Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.