Learn More
ELF5, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human ELF5 peptide using ARM Technology.
Supplier: Abnova Corporation H00002001K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene is a member of an epithelium-specific subclass of the Ets transcritpion factor family. In addition to its role in regulating the later stages of terminal differentiation of keratinocytes, it appears to regulate a number of epithelium-specific genes found in tissues containing glandular epithelium such as salivary gland and prostate. It has very low affinity to DNA due to its negative regulatory domain at the amino terminus. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
ELF5 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
ELF5 | |
ELF5 | |
A synthetic peptide of human ELF5 | |
RUO | |
2001 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
E74-like factor 5 (ets domain transcription factor) | |
ELF5 | |
ESE2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.