Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elk-1 (pS383) Mouse, Alexa Fluor 647, Clone: M21-1721, BD

Catalog No. BDB560413 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560413 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560413 Supplier BD Biosciences Supplier No. 560413
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Elk-1 is a 62kDa member of the Ets oncogene family of transcription factors and a member of the subfamily of ternary complex factors (TCF). Ets proteins mediate a variety of gene activities in response to serum and growth factors. Proteins in the TCF subfamily form a ternary complex by binding to the Serum Response Element (SRE) in conjunction with a dimer of Serum Response Factors (SRF). Elk-1 is phosphorylated by mitogen-activated protein (MAP) kinase pathways in vivo at a cluster of S/T motifs at its carboxy-terminus.
Phosphorylation at these sites, particularly at serine 383 (S383), is critical for Elk-1 transcriptional activation. Studies have shown that Elk-1 is a direct target of activated MAP kinase, and that it is also a target of the stress-activated kinase SAPK/JNK. The M21-1721 monoclonal antibody recognizes the phosphorylated S383 of activated Elk-1. The orthologous phosphorylation sites of mouse and rat Elk-1 are S384 and S382, respectively.

Host Species: Mouse
Clone: M21-1721
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Phosphorylated Human Elk-1 Peptide

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Elk-1 (pS383)
Applications Flow Cytometry
Classification Monoclonal
Clone M21-1721
Conjugate Alexa Fluor 647
Description ELK1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human Elk-1 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.