Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMAPII Rat anti-Human, Clone: M7/1, MilliporeSigma™

Catalog No. MABS278MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS278MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS278MI Supplier MilliporeSigma Supplier No. MABS278
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-EMAPII Antibody, clone M7/1, (Preservat ive free) detects level of EMAPII and has been published and validated for use in Western Blotting, Activity Assay and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMAPII
Applications Functional Assay, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone M7/1
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing PBS without preservatives.
Gene Accession No. NP_031952
Gene Symbols Aimp1, Emap2, Scye1
Host Species Rat
Immunogen His-tagged recombinant protein corresponding to mouse EMAPII.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.