Learn More
EME1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human EME1 peptide using ARM Technology.
Supplier: Abnova Corporation H00146956K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a protein that complexes with methyl methanesulfonate-sensitive UV-sensitive 81 protein to form an endonuclease complex. The encoded protein interacts with specifc DNA structures including nicked Holliday junctions, 3'-flap structures and aberrant replication fork structures. This protein may be involved in repairing DNA damage and in maintaining genomic stability
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
EME1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
EME1 | |
EME1 | |
A synthetic peptide of human EME1 | |
RUO | |
146956 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
essential meiotic endonuclease 1 homolog 1 (S. pombe) | |
EME1 | |
FLJ31364, MMS4L | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.