Learn More
EMR4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EMR4.
Supplier: Abnova Corporation H00326342A01
Description
This gene is a member of the EGF-TM7 receptor gene family which is thought to play a role in leukocyte adhesion and migration. In other vertebrates, including nonhuman primates, this gene encodes a protein containing N-terminal EGF domains and a C-terminal transmembrane domain. Sequence evidence for the human gene, however, indicates nucleotide deletion in the genomic sequence would result in frameshift and early termination of translation. A protein expressed by this gene would be soluble rather than expressed on the cell surface. As the encoded protein has not been detected, this gene may represent a transcribed pseudogene. [provided by RefSeq
Sequence: GSEAKNSGASCPPCPKYASCHNSTHCTCEDGFRARSGRTYFHDSSEKCEDINECETGLAKCKYKAYCRNKVGGSpecifications
EMR4 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant EMR4. | |
EMR4P | |
EMR4/FIRE/GPR127/PGR16 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
XM_377506 | |
EMR4P | |
EMR4 (XP_377506, 21 a.a. ∼ 93 a.a) partial recombinant protein with GST tag. | |
RUO | |
326342 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.