Learn More
Description
This gene is a member of the EGF-TM7 receptor gene family which is thought to play a role in leukocyte adhesion and migration. In other vertebrates, including nonhuman primates, this gene encodes a protein containing N-terminal EGF domains and a C-terminal transmembrane domain. Sequence evidence for the human gene, however, indicates nucleotide deletion in the genomic sequence would result in frameshift and early termination of translation. A protein expressed by this gene would be soluble rather than expressed on the cell surface. As the encoded protein has not been detected, this gene may represent a transcribed pseudogene. [provided by RefSeq
Specifications
Specifications
| Antigen | EMR4P |
| Applications | ELISA, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 1G10 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant EMR4P. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | EMR4P |
| Gene Accession No. | XM_377506 |
| Gene Alias | EMR4/FIRE/GPR127/PGR16 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.