Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

empty spiracles homeobox 2, Mouse, Clone: 4D8, Abnova™

Catalog No. 89000625 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-625 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-625 Supplier Abnova Corporation Supplier No. H00002018M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant EMX2.

This gene encodes a homeobox-containing transcription factor that is the homolog to the 'empty spiracles' gene in Drosophila. Research on this gene in humans has focused on its expression in three tissues: dorsal telencephalon, olfactory neuroepithelium, and urogenetial system. It is expressed in the dorsal telencephalon during development in a low rostral-lateral to high caudal-medial gradient and is proposed to pattern the neocortex into defined functional areas. It is also expressed in embryonic and adult olfactory neuroepithelia where it complexes with eukaryotic translation initiation factor 4E (eIF4E) and possibly regulates mRNA transport or translation. In the developing urogenital system, it is expressed in epithelial tissues and is negatively regulated by HOXA10. Alternative splicing results in multiple transcript variants encoding distinct proteins

Sequence: HSPHPLFASQQRDPSTFYPWLIHRYRYLGHRFQGNDTSPESFLLHNALARKPKRIRTAFSPSQLLRLEHAFEKNHYVVGAERKQLAHSLSLTETQVKV

Specifications

Antigen empty spiracles homeobox 2
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 4D8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant EMX2.
Formulation PBS with no preservative; pH 7.4
Gene EMX2
Gene Accession No. NM_004098
Gene Symbols EMX2
Host Species Mouse
Immunogen EMX2 (NP_004089, 103 a.a. ∼ 200 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2018
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.