Learn More
EN1, Mouse, Clone: 3E5, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant EN1.
Supplier: Abnova Corporation H00002019M02
Description
Homeobox-containing genes are thought to have a role in controlling development. In Drosophila, the 'engrailed' (en) gene plays an important role during development in segmentation, where it is required for the formation of posterior compartments. Different mutations in the mouse homologs, En1 and En2, produced different developmental defects that frequently are lethal. The human engrailed homologs 1 and 2 encode homeodomain-containing proteins and have been implicated in the control of pattern formation during development of the central nervous system. [provided by RefSeq]
Sequence: SQQPLVWPAWVYCTRYSDRPSSGPRTRKLKKKKNEKEDKRPRTAFTAEQLQRLKAEFQANRYITEQRRQTLAQELSLNESQIKIWFQNKRAKIKKATGIKNGLALHLMAQGLYNHSTTTVQDKDESE*Specifications
EN1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001426 | |
Mouse | |
Affinity chromatography | |
RUO | |
2019 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
ELISA | |
3E5 | |
Mouse monoclonal antibody raised against a full length recombinant EN1. | |
EN1 | |
EN1 | |
EN1 (NP_001417, 266 a.a. ∼ 392 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.