Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

engrailed homeobox 1, Mouse, Clone: 3H2, Abnova™

Catalog No. 89000626 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-626 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-626 Supplier Abnova Corporation Supplier No. H00002019M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant EN1.

Homeobox-containing genes are thought to have a role in controlling development. In Drosophila, the 'engrailed' (en) gene plays an important role during development in segmentation, where it is required for the formation of posterior compartments. Different mutations in the mouse homologs, En1 and En2, produced different developmental defects that frequently are lethal. The human engrailed homologs 1 and 2 encode homeodomain-containing proteins and have been implicated in the control of pattern formation during development of the central nervous system. [provided by RefSeq]

Sequence: SQQPLVWPAWVYCTRYSDRPSSGPRTRKLKKKKNEKEDKRPRTAFTAEQLQRLKAEFQANRYITEQRRQTLAQELSLNESQIKIWFQNKRAKIKKATGIKNGLALHLMAQGLYNHSTTTVQDKDESE*

Specifications

Antigen engrailed homeobox 1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 3H2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant EN1.
Formulation PBS with no preservative; pH 7.4
Gene EN1
Gene Accession No. NM_001426
Gene Symbols EN1
Host Species Mouse
Immunogen EN1 (NP_001417, 266 a.a. ∼ 392 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2019
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.