Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENAM, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89201534 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-201-534 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-201-534 Supplier Abnova Corporation Supplier No. H00010117K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human ENAM peptide using ARM Technology.

Dental enamel is a highly mineralized tissue with 85% of its volume occupied by unusually large, highly organized, hydroxyapatite crystals. This highly organized and unusual structure is thought to be rigorously controlled in ameloblasts through the interaction of a number of organic matrix molecules that include enamelin, amelogenin (AMELX; MIM 300391), ameloblastin (AMBN; MIM 601259), tuftelin (TUFT1; MIM 600087), dentine sialophosphoprotein (DSPP; MIM 125485), and a variety of enzymes. Enamelin is the largest protein in the enamel matrix of developing teeth and comprises approximately 5% of total enamel matrix protein.[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen ENAM
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description enamelin
Formulation In 1X PBS, pH 7.4
Gene ENAM
Gene Accession No. ENAM
Gene Alias ADAI, AI1C, AIH2
Gene Symbols ENAM
Host Species Rabbit
Immunogen A synthetic peptide of human ENAM
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10117
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.