Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endoglin Mouse anti-Human, Mouse, Rat, Clone: P3D1, MilliporeSigma™

Catalog No. 051424MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-142-4MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-142-4MI Supplier MilliporeSigma Supplier No. 051424
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Endoglin Antibody, clone P3D1 is an antibody against Endoglin for use in western blot, immunohistochemistry, immunocytochemistry, immunoprecipitation, flow cytometry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endoglin
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone P3D1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2a in 0.02M phosphate buffer, 0.25M sodium chloride, 0.1% sodium azide, pH 7.6
Gene Accession No. NM_000118.1
Gene Symbols ENG, ORW, CD105, HHT1, END, FLJ41744, ORW1, endoglin
Host Species Mouse
Immunogen Human resting umbilical vein endothelial cells.
Purification Method Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.