Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endorphin Β Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5028MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5028MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5028MI Supplier MilliporeSigma Supplier No. AB5028

Rabbit Polyclonal Antibody

Anti-Endorphin Antibody, β is an antibody against Endorphin for use in immunohistochemistry, RIA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endorphin β
Applications Immunohistochemistry, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin Liquid. Contains 0.1% sodium azide as a preservative.
Gene Symbols POMC; pro-ACTH-endorphin; adrenocorticotropin; beta-LPH; POC; gamma-LPH; LPH; ACTH; corticotropin; beta-endorphin; beta-MSH; alpha-MSH; pro-opiomelanocortin; proopiomelanocortin; Pro-opiomelanocortin; corticotropin-lipotrophin; NPP; MSH; met-enkephalin; CLIP; gamma-MSH
Host Species Rabbit
Immunogen Synthetic peptide (H-Thy-Gly-Gly-Phe-Met-Thr-Ser-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-Leu-Phe-Lys-Asn-Ala-Ile-Ile-Lys-Asn-Ala-Tyr-Lys-Lys-Gly-Glu-OH) from human beta-endorphin conjugated to thyroglobulin.
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000939.2; NM_001035256.1
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.