Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Endosialin/CD248 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7535
This item is not returnable.
View return policy
Description
Endosialin/CD248/TEM1 Monoclonal specifically detects Endosialin/CD248/TEM1 in Mouse samples. It is validated for Immunocytochemistry.Specifications
Endosialin/CD248/TEM1 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q91V98 | |
CD248 | |
Mouse myeloma cell line NS0-derived recombinant mouse Endosialin/CD248 Val13-Arg693 (predicted) Accession # Q91V98 | |
100 μg | |
Primary | |
Detects mouse Endosialin/CD248 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant human Endosialin/CD248 is observed. | |
Mouse | |
Purified |
Immunocytochemistry | |
458606 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
2610111G01Rik, CD164 sialomucin-like 1, CD164L1, CD248, CD248 antigen, CD248 antigen, endosialin, CD248 molecule, endosialin, endosialin, MGC119479, TEM1, TEM1MGC119478, Tumor endothelial marker 1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
57124 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction