Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endostatin Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABC60
Description
Anti-Endostatin Antibody detects level of Endostatin and has been published and validated for use in western blot, immunohistochemistry (paraffin).
Specifications
Endostatin | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Col18a1 | |
Linear peptide corresponding to the nonhelical region 11 of mouse Endostatin. | |
100 μg | |
NP_001103461 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction