Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelin 1 Receptor Mouse anti-Drosophila, Unlabeled, Clone: 16, BD

Catalog No. BDB612629 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612629 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612629 Supplier BD Biosciences Supplier No. 612629

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Endothelin-1 (ET-1) is a potent vasoconstrictor that is secreted by vascular endothelial cells. ET-1, ET-2, ET-3, and endothelin β are members of a family of related peptides. All are 21 amino acid peptides with two disulfide bridges. ET-1 and ET-2 differ by only two residues, while ET-3 differs from both by six residues. ET-1 and ET-2 are highly effective on vascular smooth muscle, while the effects of ET-3 are minimal. ET-1 and its precursor, big ET-1, are the primary forms present in the circulation. Circulating ET-1 plays a pathologic role in the etiology of hypertension and antagonists are utilized in the treatment of this disease. The ET-1 receptor is a seven-transmembrane-domain receptor of the G-protein-coupled family that signals via transient increases in intracellular Ca2+. It is primarily expressed in lung and heart and is detected at lower levels in liver, brain, muscle, and kidney. ET-1 receptor interacts with ET-1, ET-2, and ET-3. However, it has a 2-3 fold higher affinity for ET-1 than ET-2 and the affinity for ET-3 is more than 100 fold lower. Thus, ligation of the ET-1 receptor by ET-1 induces increased intracellular Ca2+ that signals to promote vasoconstriction. This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Endothelin 1 Receptor
Applications Western Blot
Classification Monoclonal
Clone 16
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Rat Endothelin 1 Receptor aa. 230-352
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Drosophila
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.