Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelin Rat anti-Human, Clone: 3G10, MilliporeSigma™

Catalog No. CBL87MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL87MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL87MI Supplier MilliporeSigma Supplier No. CBL87
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Anti-Endothelin Antibody, C-terminus, clone 3G10 is validated for use in IC for the detection of Endothelin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endothelin
Applications Immunocytochemistry
Classification Monoclonal
Clone 3G10
Conjugate Unconjugated
Formulation The monoclonal is presented at in phosphate buffered saline containing 10mM sodium azide. We recommend that each laboratory determine an optimum working titre for use in its particular application.
Gene Accession No. NM_001955.3
Gene Symbols EDN1, OTTHUMP00000039245, Preproendothelin-1, ET1, PPET1, ET-1, HDLCQ7
Host Species Rat
Purification Method Protein A purfied
Quantity 1 mg
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C for up to 1 month, −20°C in undiluted aliquots for long term
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.