Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ENG, Clone: 1E5., Abnova™

Catalog No. 89300353 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-300-353 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-300-353 Supplier Abnova Corporation Supplier No. H00002022M26A

Mouse Monoclonal Antibody

Specifications

Antigen ENG
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant ENG.
Formulation In ascites fluid
Gene Accession No. BC014271
Gene Alias CD105/END/FLJ41744/HHT1/ORW/ORW1
Gene Symbols ENG
Host Species Mouse
Immunogen ENG (AAH14271, 27 a.a. ∼ 658 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2022
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.