Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enkephalin, Met Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1975MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1975MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1975MI Supplier MilliporeSigma Supplier No. AB1975
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Enkephalin, Met Antibody is validated for use in immunohistochemistry for the detection of Enkephalin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enkephalin, Met
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum in 0.05% Sodium Azide
Gene Symbols PENK; proenkephalin
Host Species Rabbit
Immunogen Met-enkephalin, Hemocyanin conjugate.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) P01210
Target Species Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.