Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eNOS/NOS III Rabbit anti-Human, Rat, Bovine, Mouse, Canine, Guinea Pig, Pig, Polyclonal, MilliporeSigma™

Catalog No. 07520MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-520-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-520-MI Supplier MilliporeSigma Supplier No. 07520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-eNOS/NOS III Antibody, CT detects level of eNOS/NOS III and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eNOS/NOS III
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.014M phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide before the addition of glycerol to 30%. Liquid at −20°C.
Gene Symbols NOS3; cNOS; ECNOS; eNOS; EC-NOS; NOSIII
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 1181-1194 (C-QERQLRGAVPWAFD) of human eNOS/NOS III. The immunizing sequence has 10/11 identical amino acids in mouse, guinea pig, bovine, pig, rat, and dog.
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_000594
Target Species Bovine, Canine, Guinea Pig, Human, Mouse, Pig, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.