Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eNOS (pS633) Mouse anti-Human, Unlabeled, Clone: 37, BD

Catalog No. BDB612665 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612665 150 μg
BDB612664 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612665 Supplier BD Biosciences Supplier No. 612665
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nitirc oxide synthase (NOS), a cell type specific enzyme, catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical that transmits signals involved in vasorelaxation, neurotransmission, and cytoxicity. In neurons and endothelial cells, constitutive NOS (cNOS) is activated by agonists that increase intracellular Ca2+; levels and enhance calmodulin binding. Neuronal NOS (nNOS) and endothelial NOS (eNOS) have recognition sites for NADPH, FAD, FMN, and calmodulin. eNOS has a unique N-myristylation consensus sequence that may explain its membrane localization. Various protein kinases have been implicated in regulation of eNOS activity, including AMPK, PKA, PKB/Akt, PKC, and CaM Kinase II. During VEGF stimulation, eNOS is transiently phosphorylated at Ser-1177 by PKB/Akt and dephosphorylated at Thr-495. At later time points, VEGF stimulation leads to an increase in Thr-495 phosphorylation mediated by PKC and a decrease in Ser-1177 phosphorylation. In addition, Ser-633 amd Ser-1177 are phosphorylated by PKA and PKG in vitro. Thus, eNOS activity may be regulated through complex phosphorylation events mediated by multiple kinases at various phosphorylation sites.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen eNOS (pS633)
Applications Western Blot
Classification Monoclonal
Clone 37
Concentration 250μg/mL
Conjugate Unconjugated
Description Nitric Oxide Synthase
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human eNOS (pS633) Peptide
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.