Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENPP-1 Antibody, FITC, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB600816F
Description
ENPP-1 Polyclonal specifically detects ENPP-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
ENPP-1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA | |
P22413 | |
ENPP1 | |
A synthetic peptide made to a C-terminal portion of the human ENPP1 protein (between residues 900-925) [UniProt P22413] | |
Affinity Purified | |
RUO | |
Primary | |
Human. Immunogen sequence has 85% homology to mouse and rat. | |
Store at 4C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
FITC | |
PBS with 0.05% Sodium Azide | |
ectonucleotide pyrophosphatase/phosphodiesterase 1, E-NPP 1, Ly-41 antigen, M6S1NPP1, Membrane component chromosome 6 surface marker 1, membrane component, chromosome 6, surface marker 1, NPPSalkaline phosphodiesterase 1, PC1, PC-1, PCA1ARHR2, PDNP1ectonucleotide pyrophosphatase/phosphodiesterase family member 1, Phosphodiesterase I/nucleotide pyrophosphatase 1, plasma-cell membrane glycoprotein 1, Plasma-cell membrane glycoprotein PC-1 | |
Goat | |
140 kDa | |
0.1 mL | |
Lipid and Metabolism | |
5167 | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction