Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENPP7 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524848
Description
ENPP7 converts sphingomyelin to ceramide. ENPP7 also has phospholipase C activity toward palmitoyl lyso-phosphocholine. ENPP7 does not appear to have nucleotide pyrophosphatase activity.
Specifications
ENPP7 | |
Polyclonal | |
ENPP7 | |
ALK-SMase, E-NPP 7, NPP-7, alkaline sphingomyelin phosphodiesterase, alkaline sphingomyelinase, ectonucleotide pyrophosphatase/phosphodiesterase family member 7, intestinal alkaline sphingomyelinase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
339221 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q6UWV6 | |
ENPP7 | |
KLH conjugated synthetic peptide between 286-313 amino acids from the central region of human ENPP7 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction