Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enterovirus 71 Mouse anti-Human, Clone: 422-8D-4C-4D, MilliporeSigma™

Catalog No. MAB979MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB979MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB979MI Supplier MilliporeSigma Supplier No. MAB979
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Enterovirus 71 Antibody, cross-reacts with Coxsackie A16, clone 422-8D-4C-4D is validated for use in IF for the detection of Enterovirus 71.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enterovirus 71
Applications Immunofluorescence
Classification Monoclonal
Clone 422-8D-4C-4D
Conjugate Unconjugated
Formulation Ascites fluid containing no preservatives.
Host Species Mouse
Purification Method Unpurified
Quantity 50 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.