Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enterovirus Mouse anti-Human, Clone: 74-5G and 422-8D-4C-4D, MilliporeSigma™

Catalog No. MAB8430MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8430MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8430MI Supplier MilliporeSigma Supplier No. MAB8430

Mouse Monoclonal Antibody

Anti-Enterovirus Antibody, 70/71 Blend, clone 74-5G and 422-8D-4C-4D is an antibody against Enterovirus for use in IF and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enterovirus
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 74-5G and 422-8D-4C-4D
Conjugate Unconjugated
Formulation In a buffer consisting of 0.02M PB+0.25M NaCl, pH=7.6 with 0.1% Sodium Azide as a preservative.
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.