Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENTPD2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526333
Description
ENTPD2 is the type 2 enzyme of the ecto-nucleoside triphosphate diphosphohydrolase family (E-NTPDase). E-NTPDases are a family of ecto-nucleosidases that hydrolyze 5′-triphosphates. This ecto-ATPase is an integral membrane protein.
Specifications
ENTPD2 | |
Polyclonal | |
ENTPD2 | |
CD39L1, NTPDase-2, CD39 antigen-like 1, NTPDase 2, ecto-ATP diphosphohydrolase 2, ecto-ATPDase 2, ecto-ATPase 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
954 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9Y5L3 | |
ENTPD2 | |
KLH conjugated synthetic peptide between 86-113 amino acids from the N-terminal region of human ENTPD2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction