Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eosinophil Major Basic Protein Mouse anti-Human, Clone: BMK13, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma CBL419
Description
Anti-Eosinophil Major Basic Protein Antibody, clone BMK13 is an antibody against Eosinophil Major Basic Protein for use in ELISA, western blot, immunohistochemistry (paraffin).
Specifications
Eosinophil Major Basic Protein | |
Monoclonal | |
Unconjugated | |
NM_002728.4 | |
Mouse | |
Primary | |
4°C for up to 1 month, −20°C in undiluted aliquots for long term | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
BMK13 | |
The monoclonal is presented at a concentration of 100μg/ml in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular applic | |
PRG2, MBP, BMPG, MGC14537 | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction