Learn More
Eotaxin Mouse anti-Human, Biotin, Clone: 10C11, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555060
Description
The monoclonal antibody 10C11 reacts with human chemokine eotaxin. The immunogen used to generate the monoclonal antibody 10C11 was synthetic human eotaxin. This antibody is routinely tested by ELISA detection. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
ELISA, Western Blotting

Specifications
Eotaxin | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Synthetic human eotaxin | |
0.5 mg | |
Immunology | |
Human | |
IgG2b κ |
ELISA | |
10C11 | |
Biotin | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.