Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eotaxin Mouse anti-Human, Unlabeled, Clone: 3C7, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555035
Description
The monoclonal antibody 3C7 reacts with human CC chemokine eotaxin. The immunogen used to generate the monoclonal antibody 3C7 was synthetic human eotaxin.
ELISA, Western Blotting

Specifications
Eotaxin | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Synthetic human eotaxin | |
0.5 mg | |
Immunology | |
Human | |
IgG1 κ |
ELISA | |
3C7 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction