Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

erythrocyte membrane protein band 4.2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005305 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-305 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-305 Supplier Abnova Corporation Supplier No. H00002038B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human EPB42 protein.

Erythrocyte membrane protein band 4.2 is an ATP-binding protein which may regulate the association of protein 3 with ankyrin. It probably has a role in erythrocyte shape and mechanical property regulation. Mutations in the EPB42 gene are associated with recessive spherocytic elliptocytosis and recessively transmitted hereditary hemolytic anemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MGQALGIKSCDFQAARNNEEHHTKALSSRRLFVRRGQPFTIILYFRAPVRAFLPALKKVALTAQTGEQPSKINRTQATFPISSLGDRKWWSAVVEERDAQSWTISVTTPADAVIGHYSLLLQVSGRKQLLLGQFTLLFNPWNREDAVFLKNEAQRMEYLLNQNGLIYLGTADCIQAESWDFGQFEGDVIDLSLRLLSKDKQVEKWSQPVHVARVLGALLHFLKEQRVLPTPQTQATQEGALLNKRRGSVPILRQWLTGRGRPVYDGQAWVLAAVACTVLRCLGIPARVVTTFASAQGTGGRLLIDEYYNEEGLQNGEGQRGRIWKCCEDGTLELTDSNTKYVGNNISTKGVGSDRCEDITQNYKYPEGSLQEKEVLERVEKEKMEREKDNGIRPPSLETASPLYLLLKAPSSLPLRGDAQISVTLVNHSEQEKAVQLAIGVQAVHYNGVLAAKLWRKKLHLTLSANLEKIITIGLFFSNFERNPPENTFLRLTAMATHSESNLSCFAQEDIAICRPHLAIKMPEKAEQYQPLTASVSLQNSLDAPMEDCVISILGRGLIHRERSYRFRSVWPENTMCAKFQFTPTHVGLQRLTVEVDCNMFQNLTNYKSVTVVAPELSA

Specifications

Antigen erythrocyte membrane protein band 4.2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human EPB42 protein.
Formulation No additive
Gene EPB42
Gene Accession No. BC099627.1
Gene Alias MGC116735/MGC116737/PA
Gene Symbols EPB42
Host Species Mouse
Immunogen EPB42 (AAH99627.1, 1 a.a. ∼ 619 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2038
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.