Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
enhancer of polycomb homolog 1 (Drosophila), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EPC1.
Supplier: Abnova Corporation H00080314A01
Description
Sequence: KGLHLSSTTPTALVHTSPSTAGSALLQPSNITQTSSSHSALSHQVTAANSATTQVLIGNNIRLTVPSSVATVNSIAPINARHIPRTLSAVPSSALKLAAAANCQSpecifications
enhancer of polycomb homolog 1 (Drosophila) | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant EPC1. | |
EPC1 | |
DKFZp781P2312/Epl1 | |
Mouse | |
50 μL | |
Transcription Regulation | |
Primary | |
Human, Mouse, Rat | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_025209 | |
EPC1 | |
EPC1 (NP_079485, 695 a.a. ∼ 798 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
80314 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction