Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ependymin related protein 1 (zebrafish), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018504 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-504 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-504 Supplier Abnova Corporation Supplier No. H00054749D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human EPDR1 protein.

The protein encoded by this gene is a type II transmembrane protein and is similar to two families of cell adhesion molecules, the protocadherins and ependymins. This protein may play a role in calcium dependent cell adhesion. [provided by RefSeq

Sequence: MPGRAPLRTVPGALGAWLLGGLWAWTLCGLCSLGAVGAPRPCQAPQQWEGRQVMYQQSSGRNSRALLSYDGLNQRVRVLDERKALIPCKRLFEYILLYKDGVMFQIDQATKQCSKMTLTQPWDPLDIPQNSTFEDQYSIGGPQEQITVQEWSDRKSARSYETWIGIYTVKDCYPVQETFTINYSVILSTRFFDIQLGIKDPSVFTPPSTCQMAQLEKMSEDCSW

Specifications

Antigen ependymin related protein 1 (zebrafish)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human EPDR1 protein.
Formulation PBS with no preservative; pH 7.4
Gene EPDR1
Gene Accession No. BC018299.1
Gene Alias EPDR/MERP-1/MERP1/UCC1
Gene Symbols EPDR1
Host Species Rabbit
Immunogen EPDR1 (AAH18299.1, 1 a.a. ∼ 224 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54749
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.