Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eph A2/A3/A4/A5 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB2250MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2250MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2250MI Supplier MilliporeSigma Supplier No. AB2250

Rabbit Polyclonal Antibody

Detect Eph A2/A3/A4/A5 using this Anti-Eph A2/A3/A4/A5 Antibody, phospho (Tyr 596/602) validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Eph A2/A3/A4/A5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified in PBS with .07% Azide and 30% Glycerol.
Gene Symbols ECK; EphA2
Host Species Rabbit
Immunogen Synthetic Linear Peptide.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_004291.1
Target Species Human
Content And Storage −20°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.