Learn More
EPH receptor A3, Mouse, Clone: 3E9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EPHA3.
Supplier: Abnova Corporation H00002042M02
Description
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene encodes a protein that binds ephrin-A ligands. Two alternatively spliced transcript variants have been described for this gene. [provided by RefSeq
Sequence: CPFTVKNLAMFPDTVPMDSQSLVEVRGSCVNNSKEEDPPRMYCSTEGEWLVPIGKCSCNAGYEERGFMCQACRPGFYKALDGNMKCAKCPPHSSTQEDGSMNCRCENNYFRADKDPPSMACTRPPSpecifications
EPH receptor A3 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC063282 | |
EPHA3 | |
EPHA3 (AAH63282, 202 a.a. ∼ 326 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Kinases | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
3E9 | |
Mouse monoclonal antibody raised against a partial recombinant EPHA3. | |
EPHA3 | |
ETK/ETK1/HEK/HEK4/TYRO4 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
2042 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.