Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-EPHA4 Clone: 6H7,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ AP1173100UG
Description
Specifically detects EPHA4 Clone: 6H7 in Human, Mouse samples, and it is validated for ELISA, Immuno Blotting
This protein belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands.
Specifications
EPHA4 | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse | |
Purified |
ELISA, Immunoblot, Immunoblot | |
6H7 | |
In PBS, pH 7.2. | |
A recombinant polypeptide corresponding to amino acids of 887-987 human EPHA4, expressed as a GST fusion protein | |
RUO | |
Recognizes the EPHA4 protein in NIH3T3 cells. | |
−20°C | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction