Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphB3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514609
Description
This antibody is predicted to react with mouse based on sequence homology.
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB3, a member of the Tyr family of protein kinases, is a receptor for members of the ephrin-B family; it binds to both ephrin-B1 and -B2. Expression of this Type I membrane protein is ubiquitous. The protein contains putative domains for 2 fibronectin type III and 1 sterile alpha motif (SAM).
Specifications
EphB3 | |
Polyclonal | |
EPHB3 | |
Ephrin type-B receptor 3; Tyrosine-protein kinase receptor HEK-2 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
2049 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P54753 | |
EPHB3 | |
KLH conjugated synthetic peptide between 29-59 amino acids from the N-terminal region of human EphB3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction