Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPH receptor B4, Mouse, Clone: 4G11, Abnova™

Catalog No. 89000635 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-635 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-635 Supplier Abnova Corporation Supplier No. H00002050M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant EPHB4.

Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene binds to ephrin-B2 and plays an essential role in vascular development. [provided by RefSeq

Sequence: AQLTVNLTRFPETVPRELVVPVAGSCVVDAVPAPGPSPSLYCREDGQWAEQPVTGCSCAPGFEAAEGNTKCRACAQGTFKPLSGEGSCQPCPANSHSNTIGSAVCQCRVGYFRARTDPRGAPCTTP

Specifications

Antigen EPH receptor B4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4G11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant EPHB4.
Formulation PBS with no preservative; pH 7.4
Gene EPHB4
Gene Accession No. BC052804
Gene Alias HTK/MYK1/TYRO11
Gene Symbols EPHB4
Host Species Mouse
Immunogen EPHB4 (AAH52804, 198 a.a. ∼ 323 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2050
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.