Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epithelial Cell Adhesion Molecule (EpCAM) Mouse anti-Human, Clone: mab 8, MilliporeSigma™

Catalog No. MAB4444MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4444MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4444MI Supplier MilliporeSigma Supplier No. MAB4444
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Epithelial Cell Adhesion Molecule (EpCAM) Antibody, clone 8 is validated for use in flow cytometry, immunocytochemistry, western blotting for the detection of Epithelial Cell Adhesion Molecule (EpCAM).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epithelial Cell Adhesion Molecule (EpCAM)
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone mab 8
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_002345.2
Gene Symbols EPCAM, 17-1A, 323/A3, CD326, CO-17A, CO17-1A, EGP, EGP-2, EGP34, EGP40, ESA, Ep-CAM, GA733-2, HEA125, KS1/4, KSA, Ly74, M1S2, M4S1, MH99, MIC18, MK-1, MOC31, TACST-1, TACSTD1, TROP1, hEGP-2
Host Species Mouse
Immunogen HES-3 Cells (Choo, et al. (2008) Stem Cells. Jun;26(6):1454-63)
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.