Learn More
Epithelial Sodium Channel-α Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Epithelial Sodium Channel-α |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity Purified immunoglobulin. Liquid in phosphate buffered saline, pH 7.4, containing 0.1% BSA and 0.1% sodium azide. |
| Gene Symbols | SCNN1A; SCNEA; FLJ21883; ENaCa; SCNN1; Alpha-ENaC; Alpha-NaCH; ENaCalpha |
| Host Species | Rabbit |
| Immunogen | A 20 amino acid peptide sequence near the N-terminus of rat ENaC alpha (Linguela et al. 1993; Canessa et al. 1993; Snyder et al. 1994; Waldmann et al. 1995; Kreutz et al. 1997; Voilley et al. 1994; McDonald et al. 1994; Ludwig et al. 1998; Garty & Pal |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.